Assay Details
| Gene Symbol: LOC4331532 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773517.2 | XP_015629003.1 | 1 - 2 | 251 | 68 | |
| GenBank mRNA | AK102889.1 | - | 1 - 2 | 252 | 68 | |
| AY533122.1 | - | 1 - 2 | 160 | 68 | ||
More Information
Set Membership: |
|

LOC4331532
Genomic Map