Assay Details
| Gene Symbol: LOC4331833 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777660.2 | XP_015633146.1 | 1 - 2 | 393 | 76 | |
| GenBank mRNA | AK073589.1 | - | 1 - 2 | 379 | 76 | |
More Information
Set Membership: |
|

LOC4331833
Genomic Map