Assay Details
| Gene Symbol: LOC4331833 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777660.2 | XP_015633146.1 | 2 - 3 | 563 | 82 | |
| GenBank mRNA | AK073589.1 | - | 2 - 3 | 549 | 82 | |
More Information
Set Membership: |
|

LOC4331833
Genomic Map