Assay Details
| Gene Symbol: LOC4332556 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777733.2 | XP_015633219.1 | 2 - 3 | 1356 | 89 | |
| GenBank mRNA | AK243032.1 | - | 2 - 3 | 586 | 89 | |
| AK289171.1 | - | 2 - 3 | 586 | 89 | ||
More Information
Set Membership: |
|

LOC4332556
Genomic Map