Assay Details
| Gene Symbol: LOC4332585 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773131.2 | XP_015628617.1 | 1 - 2 | 173 | 149 | |
| XM_015773133.2 | XP_015628619.1 | 1 - 2 | 173 | 149 | ||
| GenBank mRNA | AK069222.1 | - | 1 - 2 | 174 | 149 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015773131.2, XM_015773133.2 transcripts.
Set Membership: |
|

LOC4332585
Genomic Map