Assay Details
| Gene Symbol: LOC4332585 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773131.2 | XP_015628617.1 | 3 - 4 | 498 | 85 | |
| XM_015773132.2 | XP_015628618.1 | 3 - 4 | 357 | 85 | ||
| XM_015773133.2 | XP_015628619.1 | 3 - 4 | 498 | 85 | ||
| GenBank mRNA | AK069222.1 | - | 3 - 4 | 499 | 85 | |
More Information
Set Membership: |
|

LOC4332585
Genomic Map