Assay Details
| Gene Symbol: LOC4332585 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773131.2 | XP_015628617.1 | 4 - 5 | 2269 | 62 | |
| XM_015773132.2 | XP_015628618.1 | 4 - 5 | 2128 | 62 | ||
| XM_015773133.2 | XP_015628619.1 | 4 - 5 | 2269 | 62 | ||
| GenBank mRNA | AK069222.1 | - | 4 - 5 | 2270 | 62 | |
More Information
Set Membership: |
|

LOC4332585
Genomic Map