Assay Details
| Gene Symbol: LOC4332585 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773133.2 | XP_015628619.1 | 5 - 6 | 2409 | 113 | |
| GenBank mRNA | AK069222.1 | - | 5 - 6 | 2410 | 113 | |
More Information
Set Membership: |
|

LOC4332585
Genomic Map