Assay Details
| Gene Symbol: LOC4332645 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775530.2 | XP_015631016.1 | 4 - 5 | 1809 | 92 | |
| XM_015775531.2 | XP_015631017.1 | 3 - 4 | 1399 | 92 | ||
| GenBank mRNA | AK060813.1 | - | 2 - 3 | 722 | 92 | |
| AK064890.1 | - | 3 - 4 | 2162 | 92 | ||
More Information
Set Membership: |
|

LOC4332645
Genomic Map