Assay Details
| Gene Symbol: LOC4332650 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777462.2 | XP_015632948.1 | 3 - 4 | 1144 | 60 | |
| GenBank mRNA | AK103334.1 | - | 3 - 4 | 1142 | 60 | |
More Information
Set Membership: |
|

LOC4332650
Genomic Map