Assay Details
| Gene Symbol: LOC4332663 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777463.2 | XP_015632949.1 | 1 - 2 | 385 | 83 | |
| GenBank mRNA | AB110220.1 | - | 1 - 2 | 405 | 83 | |
| AK101800.1 | - | 1 - 2 | 374 | 83 | ||
More Information
Set Membership: |
|

LOC4332663
Genomic Map