Assay Details
| Gene Symbol: LOC4332663 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777463.2 | XP_015632949.1 | 2 - 3 | 569 | 78 | |
| GenBank mRNA | AB110220.1 | - | 2 - 3 | 589 | 78 | |
| AK101800.1 | - | 2 - 3 | 558 | 78 | ||
More Information
Set Membership: |
|

LOC4332663
Genomic Map