Assay Details
| Gene Symbol: LOC4332663 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777463.2 | XP_015632949.1 | 3 - 4 | 735 | 65 | |
| GenBank mRNA | AB110220.1 | - | 3 - 4 | 755 | 65 | |
| AK101800.1 | - | 3 - 4 | 724 | 65 | ||
More Information
Set Membership: |
|

LOC4332663
Genomic Map