Assay Details
Gene Symbol: LOC4332682 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015776165.2 | XP_015631651.1 | 2 - 3 | 221 | 78 | |
XM_015776166.2 | XP_015631652.1 | 2 - 3 | 181 | 78 | ||
GenBank mRNA | AK073317.1 | - | 2 - 3 | 221 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
