Assay Details
| Gene Symbol: LOC4332846 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777122.2 | XP_015632608.1 | 7 - 8 | 537 | 55 | |
| XM_015777123.2 | XP_015632609.1 | 7 - 8 | 593 | 55 | ||
| GenBank mRNA | AK061662.1 | - | 7 - 8 | 539 | 55 | |
| BT129038.1 | - | 7 - 8 | 531 | 55 | ||
| BT130719.1 | - | 7 - 8 | 528 | 55 | ||
| D00999.1 | - | 7 - 8 | 509 | 55 | ||
| L36320.1 | - | 7 - 8 | 523 | 55 | ||
More Information
Set Membership: |
|

LOC4332846
Genomic Map