Assay Details
| Gene Symbol: LOC4332914 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776765.2 | XP_015632251.1 | 10 - 11 | 2257 | 63 | |
| GenBank mRNA | AK059466.1 | - | 5 - 6 | 734 | 63 | |
More Information
Set Membership: |
|

LOC4332914
Genomic Map