Assay Details
| Gene Symbol: LOC4332914 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776765.2 | XP_015632251.1 | 8 - 9 | 2025 | 79 | |
| GenBank mRNA | AK059466.1 | - | 3 - 4 | 502 | 79 | |
More Information
Set Membership: |
|

LOC4332914
Genomic Map