Assay Details
| Gene Symbol: LOC4333686 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776860.1 | XP_015632346.1 | 1 - 2 | 402 | 89 | |
| GenBank mRNA | AK069553.1 | - | 1 - 2 | 371 | 89 | |
More Information
Set Membership: |
|

LOC4333686
Genomic Map