Assay Details
| Gene Symbol: LOC4333686 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776860.1 | XP_015632346.1 | 3 - 4 | 1137 | 95 | |
| GenBank mRNA | AK059472.1 | - | 2 - 3 | 845 | 95 | |
| AK069553.1 | - | 3 - 4 | 1106 | 95 | ||
More Information
Set Membership: |
|

LOC4333686
Genomic Map