Assay Details
| Gene Symbol: LOC4333735 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777272.2 | XP_015632758.1 | 24 - 25 | 3220 | 96 | |
| GenBank mRNA | AK062164.1 | - | 5 - 6 | 427 | 96 | |
| AK065067.1 | - | 24 - 25 | 3220 | 96 | ||
More Information
Set Membership: |
|

LOC4333735
Genomic Map