Assay Details
| Gene Symbol: LOC4333735 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777272.2 | XP_015632758.1 | 4 - 5 | 493 | 144 | |
| GenBank mRNA | AK065067.1 | - | 4 - 5 | 493 | 144 | |
| AK241357.1 | - | 1168 | 144 | |||
More Information
Set Membership: |
|

LOC4333735
Genomic Map