Assay Details
| Gene Symbol: LOC4333884 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777861.2 | XP_015633347.1 | 4 - 5 | 1127 | 109 | |
| XM_015777862.2 | XP_015633348.1 | 5 - 6 | 758 | 109 | ||
| GenBank mRNA | AK060694.1 | - | 4 - 5 | 222 | 109 | |
| AK102350.1 | - | 5 - 6 | 771 | 109 | ||
More Information
Set Membership: |
|

LOC4333884
Genomic Map