Assay Details
| Gene Symbol: LOC4333891 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777393.2 | XP_015632879.1 | 2 - 3 | 529 | 64 | |
| GenBank mRNA | AK073205.1 | - | 2 - 3 | 529 | 64 | |
| AK098968.1 | - | 2 - 3 | 523 | 64 | ||
More Information
Set Membership: |
|

LOC4333891
Genomic Map