Assay Details
| Gene Symbol: LOC4333891 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777393.2 | XP_015632879.1 | 4 - 5 | 967 | 103 | |
| GenBank mRNA | AK073205.1 | - | 4 - 5 | 967 | 103 | |
| AK098968.1 | - | 4 - 5 | 961 | 103 | ||
More Information
Set Membership: |
|

LOC4333891
Genomic Map