Assay Details
| Gene Symbol: LOC4333896 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773208.2 | XP_015628694.1 | 1 - 2 | 391 | 71 | |
| GenBank mRNA | AB180689.1 | - | 1 - 2 | 245 | 71 | |
| AK063913.1 | - | 1 - 2 | 198 | 71 | ||
| AK099290.1 | - | 1 - 2 | 198 | 71 | ||
More Information
Set Membership: |
|

LOC4333896
Genomic Map