Assay Details
| Gene Symbol: LOC4333896 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015773208.2 | XP_015628694.1 | 5 - 6 | 690 | 77 | |
| GenBank mRNA | AB180689.1 | - | 5 - 6 | 544 | 77 | |
| AK063913.1 | - | 5 - 6 | 497 | 77 | ||
| AK099290.1 | - | 5 - 6 | 497 | 77 | ||
More Information
Set Membership: |
|

LOC4333896
Genomic Map