Assay Details
| Gene Symbol: LOC4333953 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772784.2 | XP_015628270.1 | 5 - 6 | 829 | 126 | |
| XM_015772785.2 | XP_015628271.1 | 5 - 6 | 827 | 126 | ||
| GenBank mRNA | AK068261.1 | - | 5 - 6 | 744 | 126 | |
| BT129118.1 | - | 3 - 4 | 443 | 126 | ||
More Information
Set Membership: |
|

LOC4333953
Genomic Map