Assay Details
| Gene Symbol: LOC4334058 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775746.2 | XP_015631232.1 | 2 - 3 | 509 | 124 | |
| GenBank mRNA | AK071772.1 | - | 2 - 3 | 504 | 124 | |
More Information
Set Membership: |
|

LOC4334058
Genomic Map