Assay Details
| Gene Symbol: LOC4330237 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770431.2 | XP_015625917.1 | 14 - 15 | 2180 | 142 | |
| XM_015770432.2 | XP_015625918.1 | 15 - 16 | 2099 | 142 | ||
| GenBank mRNA | AK066478.1 | - | 5 - 6 | 599 | 142 | |
| AK067099.1 | - | 14 - 15 | 2144 | 142 | ||
| AK103515.1 | - | 14 - 15 | 1876 | 142 | ||
More Information
Set Membership: |
|

LOC4330237
Genomic Map