Assay Details
| Gene Symbol: LOC4330237 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770431.2 | XP_015625917.1 | 24 - 25 | 3347 | 119 | |
| XM_015770432.2 | XP_015625918.1 | 25 - 26 | 3266 | 119 | ||
| GenBank mRNA | AK066478.1 | - | 15 - 16 | 1766 | 119 | |
| AK067099.1 | - | 24 - 25 | 3311 | 119 | ||
| AK103515.1 | - | 24 - 25 | 3043 | 119 | ||
More Information
Set Membership: |
|

LOC4330237
Genomic Map