Assay Details
| Gene Symbol: LOC4330237 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770431.2 | XP_015625917.1 | 27 - 28 | 3622 | 140 | |
| XM_015770432.2 | XP_015625918.1 | 28 - 29 | 3541 | 140 | ||
| GenBank mRNA | AK067099.1 | - | 27 - 28 | 3586 | 140 | |
More Information
Set Membership: |
|

LOC4330237
Genomic Map