Assay Details
| Gene Symbol: LOC4330237 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770431.2 | XP_015625917.1 | 32 - 33 | 4069 | 74 | |
| XM_015770432.2 | XP_015625918.1 | 33 - 34 | 3988 | 74 | ||
| GenBank mRNA | AK066478.1 | - | 22 - 23 | 2695 | 74 | |
More Information
Set Membership: |
|

LOC4330237
Genomic Map