Assay Details
| Gene Symbol: LOC4330237 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770431.2 | XP_015625917.1 | 6 - 7 | 1107 | 86 | |
| XM_015770432.2 | XP_015625918.1 | 7 - 8 | 1026 | 86 | ||
| GenBank mRNA | AK067099.1 | - | 6 - 7 | 1071 | 86 | |
| AK103515.1 | - | 6 - 7 | 803 | 86 | ||
More Information
Set Membership: |
|

LOC4330237
Genomic Map