Assay Details
| Gene Symbol: LOC4330302 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770978.1 | XP_015626464.1 | 5 - 6 | 1256 | 126 | |
| GenBank mRNA | AK059030.1 | - | 1 - 2 | 81 | 126 | |
| AK073916.1 | - | 5 - 6 | 1182 | 126 | ||
More Information
Set Membership: |
|

LOC4330302
Genomic Map