Assay Details
| Gene Symbol: LOC4330430 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767208.2 | XP_015622694.1 | 2 - 3 | 835 | 93 | |
| XM_015767209.2 | XP_015622695.1 | 2 - 3 | 837 | 93 | ||
| GenBank mRNA | AK102954.1 | - | 2 - 3 | 720 | 93 | |
More Information
Set Membership: |
|

LOC4330430
Genomic Map