Assay Details
| Gene Symbol: LOC4330430 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015767208.2 | XP_015622694.1 | 5 - 6 | 1903 | 87 | |
| GenBank mRNA | AK102954.1 | - | 5 - 6 | 1788 | 87 | |
More Information
Set Membership: |
|

LOC4330430
Genomic Map