Assay Details
| Gene Symbol: LOC4331068 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770164.1 | XP_015625650.1 | 12 - 13 | 2081 | 98 | |
| XM_015770165.1 | XP_015625651.1 | 12 - 13 | 2081 | 98 | ||
| GenBank mRNA | AK067109.1 | - | 12 - 13 | 2054 | 98 | |
More Information
Set Membership: |
|

LOC4331068
Genomic Map