Assay Details
| Gene Symbol: LOC4331271 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771522.2 | XP_015627008.1 | 2 - 3 | 611 | 78 | |
| XM_015771523.2 | XP_015627009.1 | 2 - 3 | 603 | 78 | ||
| XM_015771524.2 | XP_015627010.1 | 2 - 3 | 455 | 78 | ||
| GenBank mRNA | AK060083.1 | - | 2 - 3 | 463 | 78 | |
| AK099584.1 | - | 2 - 3 | 531 | 78 | ||
| AK106092.1 | - | 2 - 3 | 476 | 78 | ||
| AK106171.1 | - | 3 - 4 | 605 | 78 | ||
More Information
Set Membership: |
|

LOC4331271
Genomic Map