Assay Details
| Gene Symbol: LOC4331271 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771522.2 | XP_015627008.1 | 3 | 795 | 230 | |
| XM_015771523.2 | XP_015627009.1 | 3 | 787 | 230 | ||
| XM_015771524.2 | XP_015627010.1 | 3 | 639 | 230 | ||
| GenBank mRNA | AK060083.1 | - | 3 | 647 | 230 | |
| AK099584.1 | - | 3 | 715 | 230 | ||
| AK106092.1 | - | 3 | 660 | 230 | ||
| AK106171.1 | - | 4 - 5 | 116 | |||
More Information
Set Membership: |
|

LOC4331271
Genomic Map