Assay Details
| Gene Symbol: LOC4331327 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772615.2 | XP_015628101.1 | 4 - 5 | 856 | 147 | |
| GenBank mRNA | AK242173.1 | - | 4 - 5 | 852 | 147 | |
More Information
Set Membership: |
|

LOC4331327
Genomic Map