Assay Details
| Gene Symbol: LOC4331446 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776714.2 | XP_015632200.1 | 3 - 4 | 397 | 91 | |
| GenBank mRNA | AK062161.1 | - | 3 - 4 | 357 | 91 | |
| AK102397.1 | - | 3 - 4 | 346 | 91 | ||
| Gene Symbol: LOC4328727 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK289016.1 | - | 1958 | 91 | ||
More Information
Set Membership: |
|

LOC4331446
Genomic Map