Assay Details
| Gene Symbol: LOC9272246 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775692.2 | XP_015631178.1 | 10 - 11 | 1502 | 75 | |
| GenBank mRNA | AK063432.1 | - | 4 - 5 | 1372 | 75 | |
More Information
Set Membership: |
|

LOC9272246
Genomic Map