Assay Details
| Gene Symbol: LOC9270576 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772424.1 | XP_015627910.1 | 1 - 2 | 162 | 111 | |
| GenBank mRNA | AK072434.1 | - | 1 - 2 | 101 | 111 | |
| BT132393.1 | - | 1 - 2 | 102 | 111 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015772424.1 transcripts.
Set Membership: |
|

LOC9270576
Genomic Map