Assay Details
Gene Symbol: LOC9270576 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015772424.1 | XP_015627910.1 | 4 - 5 | 400 | 138 | |
GenBank mRNA | BT132393.1 | - | 4 - 5 | 340 | 138 |

More Information
Set Membership: |
![]() ![]() ![]() |
