Assay Details
Gene Symbol: LOC9270576 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015772424.1 | XP_015627910.1 | 5 - 6 | 474 | 65 | |
GenBank mRNA | AK072434.1 | - | 3 - 4 | 1883 | 65 | |
BT132393.1 | - | 5 - 6 | 414 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
