Assay Details
| Gene Symbol: LOC4332183 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776735.2 | XP_015632221.1 | 4 - 5 | 1312 | 113 | |
| XM_015776736.2 | XP_015632222.1 | 5 - 6 | 1442 | 113 | ||
| XM_015776738.2 | XP_015632224.1 | 5 - 6 | 1392 | 113 | ||
| XM_015776739.2 | XP_015632225.1 | 4 - 5 | 1316 | 113 | ||
| XM_015776741.2 | XP_015632227.1 | 5 - 6 | 1447 | 113 | ||
| GenBank mRNA | AK066920.1 | - | 5 - 6 | 646 | 113 | |
More Information
Set Membership: |
|

LOC4332183
Genomic Map