Assay Details
| Gene Symbol: LOC4332183 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776735.2 | XP_015632221.1 | 5 - 6 | 1873 | 98 | |
| XM_015776736.2 | XP_015632222.1 | 6 - 7 | 2003 | 98 | ||
| XM_015776738.2 | XP_015632224.1 | 6 - 7 | 1953 | 98 | ||
| XM_015776739.2 | XP_015632225.1 | 6 - 7 | 1795 | 98 | ||
| XM_015776741.2 | XP_015632227.1 | 7 - 8 | 1926 | 98 | ||
| GenBank mRNA | AK066920.1 | - | 7 - 8 | 1125 | 98 | |
More Information
Set Membership: |
|

LOC4332183
Genomic Map