Assay Details
| Gene Symbol: LOC4332290 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772765.2 | XP_015628251.1 | 1 - 2 | 158 | 124 | |
| GenBank mRNA | AK058212.1 | - | 1 - 2 | 162 | 124 | |
| AK071625.1 | - | 1 - 2 | 107 | 124 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015772765.2 transcripts.
Set Membership: |
|

LOC4332290
Genomic Map