Assay Details
| Gene Symbol: LOC4332290 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772765.2 | XP_015628251.1 | 3 - 4 | 452 | 72 | |
| GenBank mRNA | AK058212.1 | - | 3 - 4 | 456 | 72 | |
| AK071625.1 | - | 3 - 4 | 401 | 72 | ||
More Information
Set Membership: |
|

LOC4332290
Genomic Map