Assay Details
| Gene Symbol: LOC4332290 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772765.2 | XP_015628251.1 | 4 - 5 | 893 | 103 | |
| GenBank mRNA | AK058212.1 | - | 4 - 5 | 897 | 103 | |
| AK071625.1 | - | 4 - 5 | 842 | 103 | ||
More Information
Set Membership: |
|

LOC4332290
Genomic Map